(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%

Supplier: Thermo Fisher Scientific

L19689.06
ALFAL19689.06EA 0 SGD
ALFAL19689.06
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine

Purity: 98% CAS No.: 114636-30-5 Molecular Formula: C13H18N2O Formula Weight: 218.30 Storage Temperature: 2°C to 8°C Melting Point: 44-46° Beilstein Reference: 6479772 Optical Rotation: -38° (c=1 in methanol) MDL No.: MFCD03788746

Formula: C₁₃H₁₈N₂O
Melting Pt: 44…46 °C
MDL Number: MFCD03788746
CAS Number: 114636-30-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR