3-Bromo-5-methylpyridin-2-ol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Bromo-2-hydroxy-5-methylpyridine

L19849.06
ALFAL19849.06EA 0 SGD
ALFAL19849.06
3-Bromo-5-methylpyridin-2-ol ≥97%
3-Bromo-5-methylpyridin-2-ol

Purity: 97% CAS No.: 17282-02-9 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Melting Point: 156-158° Beilstein Reference: 1423374 MDL No.: MFCD03791312

Formula: C6H6BrNO
MW: 188.02 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03791312
CAS Number: 17282-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR