4-(Methylthio)benzyl chloride ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: 4-(Methylthio)benzylchloride

L19877.06
ALFAL19877.06EA 0 SGD
ALFAL19877.06
4-(Methylthio)benzyl chloride ≥99%
4-(Methylthio)benzyl chloride

Purity: 99% CAS No.: 874-87-3 Molecular Formula: C8H9ClS Formula Weight: 172.68 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6070 Boiling Point: 132°/22mm UN No.: UN3265 MDL No.: MFCD00270118

Formula: C₈H₉ClS
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00270118
CAS Number: 874-87-3
UN: 3265
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR