3'-Fluoro-4'-hydroxyacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

ALFAL19892.MDEA 0 SGD
ALFAL19892.MD
3'-Fluoro-4'-hydroxyacetophenone ≥98%
3'-Fluoro-4'-hydroxyacetophenone

Purity: 98+% CAS No.: 403-14-5 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 127-129° Beilstein Reference: 1936529

Formula: C₈H₇FO₂
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00215840
CAS Number: 403-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR