2,6-Bis(trifluoromethyl)phenylboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,6-Bis(trifluoromethyl)benzeneboronic acid

L19952.03 L19952.06 L19952.MD
ALFAL19952.03EA 232 SGD
ALFAL19952.03 ALFAL19952.06 ALFAL19952.MD
2,6-Bis(trifluoromethyl)phenylboronic acid ≥97%
2,6-Bis(trifluoromethyl)phenylboronic acid

Purity: 97% CAS No.: 681812-07-7 Molecular Formula: C8H5BF6O2 Formula Weight: 257.93 Melting Point: ca 150° MDL No.: MFCD04039321

Formula: C₈H₅BF₆O₂
Melting Pt: ∼ 150 °C
Storage Temperature: Ambient
MDL Number: MFCD04039321
CAS Number: 681812-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR