Ethylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

L19959.03 L19959.06 L19959.14
ALFAL19959.03EA 113 SGD
ALFAL19959.03 ALFAL19959.06 ALFAL19959.14
Ethylboronic acid ≥98%
Ethylboronic acid

Purity: 98% CAS No.: 4433-63-0 Molecular Formula: C2H7BO2 Formula Weight: 73.89 Storage Temperature: 2°C to 8°C Odour: Odourless Beilstein Reference: 1731546 MDL No.: MFCD01074536

Formula: C₂H₇BO₂
MW: 73.89 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074536
CAS Number: 4433-63-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR