(S)-(+)-1-(1-Naphthyl)ethyl isothiocyanate ≥97%

Supplier: Thermo Fisher Scientific
Danger

L19981.03
ALFAL19981.03EA 0 SGD
ALFAL19981.03
(S)-(+)-1-(1-Naphthyl)ethyl isothiocyanate ≥97%
(S)-(+)-1-(1-Naphthyl)ethyl isothiocyanate

Purity: 97% CAS No.: 131074-55-0 Molecular Formula: C13H11NS Formula Weight: 213.30 Storage Temperature: 2°C to 8°C Refractive Index: 1.6540 Beilstein Reference: 8682202 UN No.: UN2922 MDL No.: MFCD04039333

Formula: C₁₃H₁₁NS
Density: 1,154
MDL Number: MFCD04039333
CAS Number: 131074-55-0
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR