2-Bromo-4-iodopyridine ≥97%

Supplier: Thermo Fisher Scientific
Warning

L20015.03 L20015.06
ALFAL20015.03EA 0 SGD
ALFAL20015.03 ALFAL20015.06
2-Bromo-4-iodopyridine ≥97%
2-Bromo-4-iodopyridine

Purity: 97% CAS No.: 100523-96-4 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Melting Point: 61° Beilstein Reference: 108925

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03085768
CAS Number: 100523-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR