2,3,6-Trifluoro-4-(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Scientific
Warning

L20053.03
ALFAL20053.03EA 485 SGD
ALFAL20053.03
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine ≥97%
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 84940-46-5 Molecular Formula: C6HF6N Formula Weight: 201.07 Refractive Index: 1.3774 Boiling Point: 104-106° UN No.: 1993 MDL No.: MFCD03840203

Formula: C₆HF₆N
MW: 201,07 g/mol
Boiling Pt: 104…106 °C
Density: 1,579
Storage Temperature: Ambient
MDL Number: MFCD03840203
CAS Number: 84940-46-5
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR