Methyl-5-methyl-2-hexenoate 97%

Supplier: Thermo Fisher Scientific
Warning

L20190.03
ALFAL20190.03EA 96.1 SGD
ALFAL20190.03
Methyl-5-methyl-2-hexenoate 97%
Methyl-5-methyl-2-hexenoate

Purity: 97% CAS No.: 75513-56-3 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 57°(134°F) Refractive Index: 1.4350 Boiling Point: 95-100°/26mm Beilstein Reference: 5240071 UN No.: UN3272 MDL No.: MFCD00137613

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 95…100 °C (26 mmHg)
Flash Pt: 57 °C (134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00137613
CAS Number: 75513-56-3
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR