Cerium(IV) sulphate, anhydrous 97%

Supplier: Thermo Fisher Scientific
Danger

L20254.36
ALFAL20254.36EA 2880 SGD
ALFAL20254.36
Cerium(IV) sulphate, anhydrous 97%
Cerium (IV) sulphate

Purity: 97% CAS No.: 13590-82-4 Molecular Formula: Ce(SO4)2 Formula Weight: 332.24 Merck Reference: 14,1990 UN No.: 3085 MDL No.: MFCD00148852

Formula: Ce(SO₄)₂
Melting Pt: 195 °C
Density: 3,91
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,1990

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR