Certified thermometers
Supplier: Alla France
595.114ASC
ALLA595.114ASCEA 244 SGD
ALLA595.114ASC
Certified thermometers
Thermometers Analogue Thermometers
This thermometer is designed to measure the temperature of liquid and gaseous media in various industries, including the petrochemical, oil refining, as well as in the fuel and energy complex.
  • Yellow back
Thermometers indicate factory number, type of standard and measurement range. The temperature scale is applied with dark indelible enamel, as well as highly resistant to aggressive chemical environments. On partial immersion thermometers, the depth is indicated by a ring line.
Until 10th October 2017 it is still allowed to use mercury thermometersMore Product Information