Sampling pouches, 400 series Kapak®
Supplier: ProAmpac
400-24 400-F3-1MB 402-24
AMPA400-24EA 96.4 SGD
AMPA400-24 AMPA400-F3-1MB AMPA402-24
Sampling pouches, 400 series Kapak®
Bags Sample Bags
63 micron high barrier, crystal clear, three side seal, heat sealable polyester or foil pouches.
  • Pouches are microwaveable, boilable and freezable
  • High barrier leak proof pouches help eliminate cross contamination of dangerous substances
  • With 9 mm seals and tear notches
  • Infrared and gamma ray resistant
Typical applications: Medical, industrial, food preservation, veterinarian, health and beauty, agriculture.
Order Now
SPECIFICATIONS
Environmentally Preferable