Sampling pouches, 500 series Kapak®
Supplier: ProAmpac
504-24 503-24 500-24 508-24
AMPA504-24EA 197 SGD
AMPA504-24 AMPA503-24 AMPA500-24 AMPA508-24
Sampling pouches, 500 series Kapak®
Bags Sample Bags
114 micron high barrier, crystal clear, three side seal, heat sealable polyester pouches.
  • Pouches are microwaveable, boilable and freezable
  • High barrier leak proof pouches help eliminate cross contamination of dangerous substances
  • With 9 mm seals
  • Infrared and gamma ray resistant
  • Once sealed, these pouches can also be surface sterilised using VHP
These pouches can be used for: tissue and blood samples, formalin samples, allografts, sample retains, overwrap for medical trays and kits, medical device sterilisation - gamma irradiation, introducingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable