HPLC columns, HALO® Biphenyl
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-111 91812-211 91812-311 91812-411 91812-511 91812-611 91812-711 91812-911 91813-111 91813-211 91813-311 91813-411 91813-511 91813-611 91813-711 91813-911 92810-411 92810-611 92810-711 92811-411 92811-611 92811-711 92812-111 92812-211 92812-311 92812-411 92812-511 92812-611 92812-711 92812-911 92813-111 92813-211 92813-311 92813-411 92813-511 92813-611 92813-711 92813-911 92814-111 92814-211 92814-311 92814-411 92814-511 92814-611 92814-711 92814-911 95810-411 95810-611 95810-711 95810-911 95812-111 95812-211 95812-311 95812-411 95812-511 95812-611 95812-711 95812-911 95813-111 95813-211 95813-311 95813-411 95813-511 95813-611 95813-711 95813-911 95814-111 95814-211 95814-311 95814-411 95814-511 95814-611 95814-711 95814-911 98215-411 98215-611 98215-711 98216-411 98216-611 98216-711
AMTE91812-111EA 1510 SGD
AMTE91812-111 AMTE91812-211 AMTE91812-311 AMTE91812-411 AMTE91812-511 AMTE91812-611 AMTE91812-711 AMTE91812-911 AMTE91813-111 AMTE91813-211 AMTE91813-311 AMTE91813-411 AMTE91813-511 AMTE91813-611 AMTE91813-711 AMTE91813-911 AMTE92810-411 AMTE92810-611 AMTE92810-711 AMTE92811-411 AMTE92811-611 AMTE92811-711 AMTE92812-111 AMTE92812-211 AMTE92812-311 AMTE92812-411 AMTE92812-511 AMTE92812-611 AMTE92812-711 AMTE92812-911 AMTE92813-111 AMTE92813-211 AMTE92813-311 AMTE92813-411 AMTE92813-511 AMTE92813-611 AMTE92813-711 AMTE92813-911 AMTE92814-111 AMTE92814-211 AMTE92814-311 AMTE92814-411 AMTE92814-511 AMTE92814-611 AMTE92814-711 AMTE92814-911 AMTE95810-411 AMTE95810-611 AMTE95810-711 AMTE95810-911 AMTE95812-111 AMTE95812-211 AMTE95812-311 AMTE95812-411 AMTE95812-511 AMTE95812-611 AMTE95812-711 AMTE95812-911 AMTE95813-111 AMTE95813-211 AMTE95813-311 AMTE95813-411 AMTE95813-511 AMTE95813-611 AMTE95813-711 AMTE95813-911 AMTE95814-111 AMTE95814-211 AMTE95814-311 AMTE95814-411 AMTE95814-511 AMTE95814-611 AMTE95814-711 AMTE95814-911 AMTE98215-411 AMTE98215-611 AMTE98215-711 AMTE98216-411 AMTE98216-611 AMTE98216-711
HPLC columns, HALO® Biphenyl
Chromatography Columns
HALO® Biphenyl columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å biphenyl columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for aromatic (pi-pi) compounds
  • Alternate selectivity to alkyl phases
  • 100% aqueous compatible
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
More Product Information