HPLC columns, HALO® HILIC
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-101 91812-201 91812-301 91812-401 91812-501 91812-601 91812-701 91812-901 91813-101 91813-201 91813-301 91813-401 91813-501 91813-601 91813-701 91813-901 92810-401 92810-601 92810-701 92811-401 92811-601 92811-701 92812-101 92812-201 92812-301 92812-401 92812-501 92812-601 92812-701 92812-901 92813-101 92813-201 92813-301 92813-401 92813-501 92813-601 92813-701 92813-901 92814-101 92814-201 92814-301 92814-401 92814-501 92814-601 92814-701 92814-901 95810-401 95810-601 95810-701 95810-901 95812-101 95812-201 95812-301 95812-401 95812-501 95812-601 95812-701 95812-901 95813-101 95813-201 95813-301 95813-401 95813-501 95813-601 95813-701 95813-901 95814-101 95814-201 95814-301 95814-401 95814-501 95814-601 95814-701 95814-901 98215-401 98215-601 98215-701 98216-401 98216-601 98216-701
AMTE91812-101EA 1510 SGD
AMTE91812-101 AMTE91812-201 AMTE91812-301 AMTE91812-401 AMTE91812-501 AMTE91812-601 AMTE91812-701 AMTE91812-901 AMTE91813-101 AMTE91813-201 AMTE91813-301 AMTE91813-401 AMTE91813-501 AMTE91813-601 AMTE91813-701 AMTE91813-901 AMTE92810-401 AMTE92810-601 AMTE92810-701 AMTE92811-401 AMTE92811-601 AMTE92811-701 AMTE92812-101 AMTE92812-201 AMTE92812-301 AMTE92812-401 AMTE92812-501 AMTE92812-601 AMTE92812-701 AMTE92812-901 AMTE92813-101 AMTE92813-201 AMTE92813-301 AMTE92813-401 AMTE92813-501 AMTE92813-601 AMTE92813-701 AMTE92813-901 AMTE92814-101 AMTE92814-201 AMTE92814-301 AMTE92814-401 AMTE92814-501 AMTE92814-601 AMTE92814-701 AMTE92814-901 AMTE95810-401 AMTE95810-601 AMTE95810-701 AMTE95810-901 AMTE95812-101 AMTE95812-201 AMTE95812-301 AMTE95812-401 AMTE95812-501 AMTE95812-601 AMTE95812-701 AMTE95812-901 AMTE95813-101 AMTE95813-201 AMTE95813-301 AMTE95813-401 AMTE95813-501 AMTE95813-601 AMTE95813-701 AMTE95813-901 AMTE95814-101 AMTE95814-201 AMTE95814-301 AMTE95814-401 AMTE95814-501 AMTE95814-601 AMTE95814-701 AMTE95814-901 AMTE98215-401 AMTE98215-601 AMTE98215-701 AMTE98216-401 AMTE98216-601 AMTE98216-701
HPLC columns, HALO® HILIC
Chromatography Columns
HALO® HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for polar analytes
  • Alternate mode to reversed-phase modes
  • Can be used in HILIC and normal-phase modes
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
pH range 1 - 8
Temperature limit 60 °C (pH 1); 40 °C (pH 8)
USP code L3