HPLC columns, HALO® C30
Supplier: ADVANCED MATERIALS TECHNOLOGY
91215-402 91215-602 91215-702 91216-402 91216-602 91216-702 92110-402 92110-602 92110-702 92111-402 92111-602 92111-702 92112-130 92112-230 92112-330 92112-430 92112-530 92112-630 92112-730 92112-930 92113-130 92113-230 92113-330 92113-430 92113-530 92113-630 92113-730 92113-930 92114-130 92114-230 92114-330 92114-430 92114-530 92114-630 92114-730 92114-930
AMTE91215-402EA 2400 SGD
AMTE91215-402 AMTE91215-602 AMTE91215-702 AMTE91216-402 AMTE91216-602 AMTE91216-702 AMTE92110-402 AMTE92110-602 AMTE92110-702 AMTE92111-402 AMTE92111-602 AMTE92111-702 AMTE92112-130 AMTE92112-230 AMTE92112-330 AMTE92112-430 AMTE92112-530 AMTE92112-630 AMTE92112-730 AMTE92112-930 AMTE92113-130 AMTE92113-230 AMTE92113-330 AMTE92113-430 AMTE92113-530 AMTE92113-630 AMTE92113-730 AMTE92113-930 AMTE92114-130 AMTE92114-230 AMTE92114-330 AMTE92114-430 AMTE92114-530 AMTE92114-630 AMTE92114-730 AMTE92114-930
HPLC columns, HALO® C30
Chromatography Columns
HALO® C30 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in 2,7 µm particle size and a wide range of column dimensions, HALO® 160 Å C30 columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for hydrophobic, long chain, structurally related isomers
  • Alternate alkyl phase with high shape selectivity
  • 100% aqueous compatible
  • Particle size: 2,7 μm superficially porous particle silica ('solid-core',
More Product Information