HPLC columns, HALO® C4, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
92710-414 92710-614 92710-714 92711-414 92711-614 92711-714 92712-114 92712-214 92712-314 92712-414 92712-514 92712-614 92712-714 92712-914 92713-114 92713-214 92713-314 92713-414 92713-514 92713-614 92713-714 92713-914 92714-114 92714-214 92714-314 92714-414 92714-514 92714-614 92714-714 92714-914 97215-414 97215-614 97215-714 97216-414 97216-614 97216-714
AMTE92710-414EA 5610 SGD
AMTE92710-414 AMTE92710-614 AMTE92710-714 AMTE92711-414 AMTE92711-614 AMTE92711-714 AMTE92712-114 AMTE92712-214 AMTE92712-314 AMTE92712-414 AMTE92712-514 AMTE92712-614 AMTE92712-714 AMTE92712-914 AMTE92713-114 AMTE92713-214 AMTE92713-314 AMTE92713-414 AMTE92713-514 AMTE92713-614 AMTE92713-714 AMTE92713-914 AMTE92714-114 AMTE92714-214 AMTE92714-314 AMTE92714-414 AMTE92714-514 AMTE92714-614 AMTE92714-714 AMTE92714-914 AMTE97215-414 AMTE97215-614 AMTE97215-714 AMTE97216-414 AMTE97216-614 AMTE97216-714
HPLC columns, HALO® C4, BIOCLASS
Chromatography Columns
HALO® BIOCLASS C4 1000 Å Protein columns made with innovative large pore Fused-Core® particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 1000 Å columns are optimised for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
  • Ideal for mAbs, ADC's, and and large proteins due to unrestricted access to the bonded phase
  • Best in class resolution for full
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Endcapped Yes
pH range 2 - 9
Temperature limit 90 °C (pH 2); 40 °C (pH 9)
USP code L26