HPLC columns, HALO® C4, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
93410-414 93410-614 93410-714 93411-414 93411-614 93411-714 93412-114 93412-214 93412-314 93412-414 93412-514 93412-614 93412-714 93412-914 93413-114 93413-214 93413-314 93413-414 93413-514 93413-614 93413-714 93413-914 93414-114 93414-214 93414-314 93414-414 93414-514 93414-614 93414-714 93414-914 94315-414 94315-614 94315-714 94316-414 94316-614 94316-714
AMTE93410-414EA 5610 SGD
AMTE93410-414 AMTE93410-614 AMTE93410-714 AMTE93411-414 AMTE93411-614 AMTE93411-714 AMTE93412-114 AMTE93412-214 AMTE93412-314 AMTE93412-414 AMTE93412-514 AMTE93412-614 AMTE93412-714 AMTE93412-914 AMTE93413-114 AMTE93413-214 AMTE93413-314 AMTE93413-414 AMTE93413-514 AMTE93413-614 AMTE93413-714 AMTE93413-914 AMTE93414-114 AMTE93414-214 AMTE93414-314 AMTE93414-414 AMTE93414-514 AMTE93414-614 AMTE93414-714 AMTE93414-914 AMTE94315-414 AMTE94315-614 AMTE94315-714 AMTE94316-414 AMTE94316-614 AMTE94316-714
HPLC columns, HALO® C4, BIOCLASS
Chromatography Columns
HALO® BIOCLASS C4 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. These columns are optimised for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
  • Ideal for large protein analysis such as mAbs and polypeptides
  • Well suited for high speed protein separations
  • Outstanding low bleed performance and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Endcapped Yes
pH range 2 - 9
Temperature limit 90 ⁰C (pH 2); 40 ⁰C (pH 9)
USP code L26