HPLC columns, HALO® Penta-HILIC
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-105 91812-205 91812-305 91812-405 91812-505 91812-605 91812-705 91812-905 91813-105 91813-205 91813-305 91813-405 91813-505 91813-605 91813-705 91813-905 92810-405 92810-605 92810-705 92811-405 92811-605 92811-705 92812-105 92812-205 92812-305 92812-405 92812-505 92812-605 92812-705 92812-905 92813-105 92813-205 92813-305 92813-405 92813-505 92813-605 92813-705 92813-905 92814-105 92814-205 92814-305 92814-405 92814-505 92814-605 92814-705 92814-905 95810-405 95810-605 95810-705 95810-905 95812-105 95812-205 95812-305 95812-405 95812-505 95812-605 95812-705 95812-905 95813-105 95813-205 95813-305 95813-405 95813-505 95813-605 95813-705 95813-905 95814-105 95814-205 95814-305 95814-405 95814-505 95814-605 95814-705 95814-905 98215-405 98215-605 98215-705 98216-405 98216-605 98216-705
AMTE91812-105EA 1510 SGD
AMTE91812-105 AMTE91812-205 AMTE91812-305 AMTE91812-405 AMTE91812-505 AMTE91812-605 AMTE91812-705 AMTE91812-905 AMTE91813-105 AMTE91813-205 AMTE91813-305 AMTE91813-405 AMTE91813-505 AMTE91813-605 AMTE91813-705 AMTE91813-905 AMTE92810-405 AMTE92810-605 AMTE92810-705 AMTE92811-405 AMTE92811-605 AMTE92811-705 AMTE92812-105 AMTE92812-205 AMTE92812-305 AMTE92812-405 AMTE92812-505 AMTE92812-605 AMTE92812-705 AMTE92812-905 AMTE92813-105 AMTE92813-205 AMTE92813-305 AMTE92813-405 AMTE92813-505 AMTE92813-605 AMTE92813-705 AMTE92813-905 AMTE92814-105 AMTE92814-205 AMTE92814-305 AMTE92814-405 AMTE92814-505 AMTE92814-605 AMTE92814-705 AMTE92814-905 AMTE95810-405 AMTE95810-605 AMTE95810-705 AMTE95810-905 AMTE95812-105 AMTE95812-205 AMTE95812-305 AMTE95812-405 AMTE95812-505 AMTE95812-605 AMTE95812-705 AMTE95812-905 AMTE95813-105 AMTE95813-205 AMTE95813-305 AMTE95813-405 AMTE95813-505 AMTE95813-605 AMTE95813-705 AMTE95813-905 AMTE95814-105 AMTE95814-205 AMTE95814-305 AMTE95814-405 AMTE95814-505 AMTE95814-605 AMTE95814-705 AMTE95814-905 AMTE98215-405 AMTE98215-605 AMTE98215-705 AMTE98216-405 AMTE98216-605 AMTE98216-705
HPLC columns, HALO® Penta-HILIC
Chromatography Columns
HALO® Penta-HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å Penta-HILIC columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for polar compounds poorly retained in reversed-phase modes
  • Alternate selectivity for HILIC mode
  • Excellent peak shape for basic compounds in HILIC mode
  • Particle sizes: 2, 2,7 or 5 μm
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Endcapped No
pH range 2 - 9
Temperature limit 60 ⁰C (pH 2); 40 ⁰C (pH 9)
USP code L95