Sleeve protector, cut resistant, HyFlex® 11-200
Supplier: Ansell
11-200
ANSE11-200EA 564 SGD
ANSE11-200
Sleeve protector, cut resistant, HyFlex® 11-200
Workwear
This high visibility sleeve with Level 5 cut performance and Level 1 thermal protection is knitted from polyester, nylon and glass fibres. The anatomic design with adjustable forearm area enables freedom of movement so workers can do the job without constraints, increasing productivity.
  • Most comfortable sleeve in the Ansell range: Generous space in the arm area reduces sweat and itchiness
  • Better abrasion resistance (Level 2) and washability mean HyFlex® 11-200 offers improved total cost of ownership
  • Due to its ergonomic design and fixing system, worker
More Product Information