High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
Supplier: Ansell
11-202
ANSE11-202EA 623 SGD
ANSE11-202
High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
Workwear
This sleeve made of polyester, nylon and glass fibre has a level EN CUT 5 performance and an increased abrasion protection. Safety is also enhanced through its visibility colour and level 1 thermal protection.
  • Sleeve delivers the best comfort through its anatomical design
  • Offers an improved fit thanks to its adjustable VELCRO® strap system
  • Freedom of movement leads to increased productivity and worker acceptance
Ideal for use in body shops (excluding welding), for glass and metal sheet handling and for stamping.
Gauge: 13
Certifications: EN 388 (More Product Information