Chemical resistant gloves, Sol-Vex®
Supplier: Ansell
117209
ANSE117209EA 295 SGD
ANSE117209
Chemical resistant gloves, Sol-Vex®
Gloves
These chemical resistant gloves are suitable for chemical processing, food processing, aerospace and automotive degreasing, machining operations using cutting oil and coolants, electronics, and other applications. Made of tough nitrile compound they provide long wear and good protection against abrasion, cuts, snags, and punctures.
  • Curved finger design and contoured palm help to reduce fatigue
  • Full cut allows for easy donning and doffing
  • Embossed texture improves grip for easier handling of wet objects
  • Strong resistance to degradation ensures that gloves
More Product Information