Heat resistant gloves, ThermaPrene™
Supplier: Ansell
214022
ANSE214022EA 131 SGD
ANSE214022
Heat resistant gloves, ThermaPrene™
Gloves
These gloves combines the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner.
  • For intermittent handling of hot objects up to 200 °C
  • Unaffected by grease and oil
  • Gauntlet cuff for additional forearm protection
  • Rough finish for good grip
  • Removable liner and integrated hanging loop allows gloves to be stored and dried quickly
Ideal for use in bakeries and canteens. Also suitable for cleaning fryers and grills, for acidic, aqueous and alcoholic food contact, for handling chilled or refrigeratedMore Product Information