Chemical resistant gloves, AlphaTec® 15-554
Supplier: Ansell
214308
ANSE214308EA 101 SGD
ANSE214308
Chemical resistant gloves, AlphaTec® 15-554
Gloves
These comfortable anatomical gloves with pre-curved fingers and winged thumb are made from a PVA compound, which remains inert in the strongest organic, aromatic and chlorinated solvents.
  • Polyvinyl alcohol coating lasts 10 - 15 times longer than any other coating
  • Soft two-piece knitted liner cushions the hand and absorbs perspiration
  • Virtually no absorption or extraction of ions
  • Exceptionally light and flexible
Length: 355 mm
Colour: Red
EN 374 (BCD), EN 388 (3121), EN 1149, Cat. III; AQL 1.0
Certifications: EN ISO 374-1:2016 Type A (More Product Information