Sleeve protectors, cut resistant
Supplier: Ansell
70-114 70-118 70-123 70-110
ANSE70-114EA 18.6 SGD
ANSE70-114 ANSE70-118 ANSE70-123 ANSE70-110
Sleeve protectors, cut resistant
Workwear
These knitted liners are constructed from 100% Kevlar® para-aramid short fibres.
  • Exceptional cut resistance with dexterity
  • Most models feature thumb slots, protecting the wrist area
They are available in different versions and lengths. All sleeves provide superior mechanical protection and cut resistance.
Gauge: 24
Certifications: EN 388 (134X), EN 407 (X1XXXX); Cat. II
Order Now
SPECIFICATIONS
Garment application Mechanical protection
Environmentally Preferable
Risk area Cuts
Gender Unisex
Fabric 100% Kevlar® para-aramid