Apron, Vinyl
Supplier: Ansell
950191_U 950190 950191
ANSE950191_UEA 777 SGD
ANSE950191_U ANSE950190 ANSE950191
Apron, Vinyl
Disposable Garments
This disposable, chemical resistant apron is made of virgin vinyl. It provides excellent protection when handling acids and caustic solutions.
  • Heat-sealed binding at the neck and bottom eliminates leaks caused by exposed elastic and thread
  • Back closure consists of two plastic snaps and nylon ties
  • No exposed elastic or metal parts to create static and corrosion
  • Full-length sleeves provide additional coverage
Ideal for use in chemical processing, oil refining, petrochemicals, beef/pork/poultry/fish processing, janitorial and maintenance work and for useMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Chemical protection
Thickness 0,2 mm