Aprons, PVC
Supplier: Ansell
56-100 PVC-45W
ANSE56-100EA 22.6 SGD
ANSE56-100 ANSEPVC-45W
Aprons, PVC
Workwear
Durable apron made from strong PVC; with nylon neck band.
  • High resistance to oils, greases and certain chemicals
  • Easy to clean
Thickness: 0,51 mm.
Certifications: EN 14605; Cat. III
Order Now
SPECIFICATIONS
Garment application Chemical protection,Oils, fats and fuels
Environmentally Preferable
Risk area
Gender Unisex
Fabric PVC
Thickness 0,51 mm