Shoe covers, CPE
Supplier: Ansell
SC-36BCPE
ANSESC-36BCPEEA 6.1 SGD
ANSESC-36BCPE
Shoe covers, CPE
Disposable Garments
Elasticated shoe covers made from chlorinated PE.
  • Protects outdoor shoes from getting wet
  • Helps prevent dirt from outdoor shoes being brought inside
Thickness: 0,038 mm
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Laboratory,Food