Apron with sleeves, MICROGARD® 2000, model 214
Supplier: Ansell
WH20B-00214-07
ANSEWH20B-00214-07EA 1310 SGD
ANSEWH20B-00214-07
Apron with sleeves, MICROGARD® 2000, model 214
Disposable Garments
These aprons are made from a microporous PE film with a non woven inner layer. The fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
  • Can be used as a hospital gown
  • Rear Velcro fastening
  • Elasticated wrists
  • Bound seams