ESD boot covers, MICROGARD® 2000, model 407
Supplier: Ansell
WH20B-00407-00
ANSEWH20B-00407-00EA 1840 SGD
ANSEWH20B-00407-00
ESD boot covers, MICROGARD® 2000, model 407
Disposable Garments
These ESD overboots are made from a microporous polyethylene film with a non woven inner layer.
  • Tie fastening
  • Elastic to top of boot
  • Bound seams
  • ESD PVC sole
  • Fit shoe size 42 - 46