Surgical undergloves, latex, sterile, Gammex® Latex Underglove
Supplier: ANSELL MEDICAL PRODUCTS
330050055 330050060 330050065 330050080 330050075 330050085 330050090
ANSM330050055EA 233 SGD
ANSM330050055 ANSM330050060 ANSM330050065 ANSM330050080 ANSM330050075 ANSM330050085 ANSM330050090
Surgical undergloves, latex, sterile, Gammex® Latex Underglove
Gloves
Double gloving is probably one of the most effective methods to reduce exposure to blood-borne pathogens, as it reduces by a factor of 10 the number of potential exposures. Double gloving also significantly reduces the perforation rate of the inner glove, and it is especially efficient with the green colour of the GAMMEX® Latex Underglove, which provides a high-strength barrier to infection with great tear resistance. This provides an effective shield against viral penetration, while at the same time a tough inner coating withstands alcohol-based disinfectants.
  • Powder-free, micro-
More Product Information
Order Now
SPECIFICATIONS
Model Underglove
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Disposable
Latex / latex-free Latex
Powdered / powder-free Powder-free
Finish Micro-textured
Cuff style Straight
EN norm EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS