Surgical gloves, polyisoprene, sterile, Gammex® Non-Latex PI Micro
Supplier: ANSELL MEDICAL PRODUCTS
340057060 340057065 340057070 340057075 340057080 340057085
ANSM340057060EA 336 SGD
ANSM340057060 ANSM340057065 ANSM340057070 ANSM340057075 ANSM340057080 ANSM340057085
Surgical gloves, polyisoprene, sterile, Gammex® Non-Latex PI Micro
Gloves
These thin gloves made from synthetic polyisoprene offer an optimal level of sensitivity, comfort, and protection against natural rubber latex allergies.
  • Powder-free, micro-textured finish
  • DPG-free polyisoprene option for healthcare workers suffering from latex allergies and chemical allergies
  • White colour makes it ideal as a double-gloving solution, together with the green GAMMEX® Non-Latex PI underglove
  • Beaded cuffs with adhesive band
  • External surface: Micro-textured and chlorinated and siliconised; internal surface: PU coated and siliconised
Order Now
SPECIFICATIONS
Model Non-Latex PI Micro
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Disposable
Latex / latex-free Latex-free
Powdered / powder-free Powder-free
Finish Micro-textured
Cuff style Beaded
EN norm EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1