Guanidinium hydrochloride 8 mol/l in aqueous solution

Supplier: PanReac AppliChem
Warning

A0860.0500
APLIA0860.0500EA 252 SGD
APLIA0860.0500
Guanidinium hydrochloride 8 mol/l in aqueous solution
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR