D(-)-Luciferin sodium salt ≥98.5%

Supplier: PanReac AppliChem

Synonyms: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt

A1006.0010
APLIA1006.0010EA 316 SGD
APLIA1006.0010
D(-)-Luciferin sodium salt ≥98.5%
D(-)-Luciferin sodium salt
Formula: C₁₁H₇N₂NaO₃S₂
MDL Number: MFCD00044938
CAS Number: 103404-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR