N-Octyl-β-D-glucopyranoside

Supplier: PanReac AppliChem
Warning

A1010.0025
APLIA1010.0025EA 2630 SGD
APLIA1010.0025
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
Melting Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8
EINECS: 249-887-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR