L(+)-Cysteine

Supplier: PanReac AppliChem
Warning

A1425.1000
APLIA1425.1000EA 5890 SGD
APLIA1425.1000
L(+)-Cysteine
L-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1,523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR