Carbenicillin disodium salt

Supplier: PanReac AppliChem
Danger

Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

A1491.0005 A1491.0010
APLIA1491.0005EA 423 SGD
APLIA1491.0005 APLIA1491.0010
Carbenicillin disodium salt
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR