Guanidinium hydrochloride

Supplier: PanReac AppliChem
Warning

A1499.1000
APLIA1499.1000EA 529 SGD
APLIA1499.1000
Guanidinium hydrochloride
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Density: 1,354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR