Amphotericin B

Supplier: PanReac AppliChem
Warning

Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

A1907.0050 A1907.0001
APLIA1907.0050EA 0 SGD
APLIA1907.0050 APLIA1907.0001
Amphotericin B
Amphotericin B
Formula: C₄₇H₇₃NO₁₇
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3
EINECS: 215-742-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR