Chloroform : isoamyl alcohol (24:1) (v/v)

Supplier: PanReac AppliChem
Warning

A1935.0500
APLIA1935.0500EA 189 SGD
APLIA1935.0500
Chloroform : isoamyl alcohol (24:1) (v/v)
Chloroform : isoamyl alcohol
Formula: CHCl₃,C₅H₁₂O
Storage Temperature: Ambient
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR