G418 disulphate

Supplier: PanReac AppliChem
Warning

A2167.0001
APLIA2167.0001EA 423 SGD
APLIA2167.0001
G418 disulphate
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR