Polyvinylpyrrolidone (K15), BioChemica®

Supplier: PanReac AppliChem

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

A2258.1000
APLIA2258.1000EA 584 SGD
APLIA2258.1000
Polyvinylpyrrolidone (K15), BioChemica®
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now

Specification Test Results

Assay (from N) 12.0 - 12.8 %
pH (5 %; H2O) 3.0 - 5.0
Heavy metals (as Pb) max. 0.001 %
Water (K.F.) max. 5 %
Solubility (10 %; H2O) clear, colorless to yellowish


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR