Dextran sulphate sodium salt, M.W. ~ 40,000

Supplier: PanReac AppliChem

A3261.0250
APLIA3261.0250EA 2260 SGD
APLIA3261.0250
Dextran sulphate sodium salt, M.W. ~ 40,000
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR