Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution

Supplier: PanReac AppliChem
Danger

Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis

A3658.1000
APLIA3658.1000EA 533 SGD
APLIA3658.1000
Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution
Acrylamide-Bis (32:1)
Density: 1,03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR