Sodium pyruvate

Supplier: PanReac AppliChem

A4859.0100 A4859.1000
APLIA4859.0100EA 309 SGD
APLIA4859.0100 APLIA4859.1000
Sodium pyruvate
Sodium pyruvate
Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1,718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR