Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution for molecular biology

Supplier: PanReac AppliChem

A5324.0001
APLIA5324.0001EA 89.8 SGD
APLIA5324.0001
Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution for molecular biology
Magnesium chloride
Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR