Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology

Supplier: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

A7006.1000
APLIA7006.1000EA 250 SGD
APLIA7006.1000
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

DNases/RNases/Proteases not detectable
Composition NaCl: 292.2 g/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR