Cytochalasin B

Supplier: PanReac AppliChem

Synonyms: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

A7657.0005
APLIA7657.0005EA 867 SGD
APLIA7657.0005
Cytochalasin B
Cytochalasin B
Formula: C₂₉H₃₇NO₅
Melting Pt: 218…221 °C
MDL Number: MFCD00077704
CAS Number: 14930-96-2
EINECS: 239-000-2
UN: 1544
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR