Patent blue V calcium salt ≥85% (by titrimetric analysis)

Supplier: PanReac AppliChem

Synonyms: Bis[hydrogen [4-[4-(diethylamino)-5'-hydroxy-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium], calcium salt, Acid Blue 3 Calcium Salt

A8187.0025
APLIA8187.0025EA 301 SGD
APLIA8187.0025
Patent blue V calcium salt ≥85% (by titrimetric analysis)
Patent blue V calcium salt
Formula: C₅₄H₆₂CaN₄O₁₄S₄
Storage Temperature: Ambient
MDL Number: MFCD00038746
CAS Number: 3536-49-0

Order Now

Specification Test Results

Assay (titr.) Min. 85% 
In H₂O insoluble matter Max. 0,20% 
Unsulphonated primary aromatic amine Max. 0,01% 
Heavy metals (as Pb) Max. 0,004% 
Subsidiary dyes Max. 2,0% 
Arsenic (As) Max. 0,0003% 
Cadmium (Cd) Max. 0,0001% 
Mercury (Hg) Max. 0,0001% 

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR